Learn More
Chloroform, 99+%, extra pure, stabilized with ethanol
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -63.0°C |
| Decomposition Information | 290°C |
| Density | 1.4800g/mL |
| Quantity | 5 L |
| Packaging | Plastic Drum |
| Physical Form | Liquid |
| Refractive Index | 1.4443 to 1.4463 |
| Assay Percent Range | 99+% |
| Residue after Evaporation | 0.002% max. |
| Specific Gravity | 1.48 |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled. |
| Precautionary Statement | GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.