Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, ACS, 99.8%, MilliporeSigma™

Catalog No. 50947956 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-956 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-947-956 Supplier MilliporeSigma Supplier No. CX1055P4
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point −63°C
Quantity 4 L
Packaging Glass Bottle (Pre-packed)
Residue after Evaporation 0.001% max
Assay Percent Range 99.8% min
Vapor Pressure 211 hPa (20°C)
Boiling Point 61°C (1013 hPa)
Solubility Information 8.7 g/L
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.