Learn More
Chloroform, ACS Grade, ≥ 99.8%, LabChem™

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Melting Point | -64°C |
| Color | Colorless |
| CAS Max % | 99, 1 |
| Decomposition Information | Chlorine |
| Density | 1,490kg/m3 |
| Quantity | 4 L |
| Identification | Passes Test |
| Packaging | Amber Glass |
| Physical Form | Liquid |
| Residue after Evaporation | ≤0.001% |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Toxic if inhaled. Causes skin and serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs (liver, kidneys) through prolonged or repeated exposure. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Do not breathe vapors. Do not eat, drink or smoke when using this product. Use only outdoors or in a well-ventilated area. Wear eye protection, face protection, protective clothing, protective gloves. Wash exposed skin thoroughly after handling. If exposed or concerned: Get medical advice/attention. If swallowed: Rinse mouth. Call a poison center or doctor if you feel unwell. If on skin: Wash with plenty of soap and water. Take off contaminated clothing and wash it before reuse. If skin irritation occurs: Get medical attention. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center or doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical attention. Store locked up in a well-ventilated place. Keep container tightly closed. Dispose of contents/container to comply with local, state and federal regulations. |
| EINECSNumber | 200-663-8 |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.