Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RSOC0020500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOC0020500 500 mL Amber Glass Bottle
RSOC00201C 1 L Amber Glass Bottle
RSOC00204C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RSOC0020500 Supplier Ricca Chemical Company Supplier No. RSOC0020500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For General Use

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Color Yellow
CAS Min % 98.0
CAS Max % 100.0
Quantity 500 mL
Packaging Amber Glass Bottle
Physical Form Liquid
Boiling Point 61°C
Percent Purity >99.8%
Grade ACS
Chemical Name or Material Chloroform
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.