Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, contains amylene, >99.9%, Solstice

Catalog No. 6000408 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-004-08 4 x 4 L Poly-coated Bottle
60-004-07 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-004-08 Supplier Solstice Supplier No. AH0494PC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-66-3 , 513-35-9
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

CAS Max % 99.9900%, <0.0100%
Density 1.49 g/cm3
Quantity 4 x 4 L
Packaging Poly-coated Bottle
UN Number UN1888
Water 0.02% max.
Beilstein 1731042
Boiling Point 62°C
Solubility Information 81.5% in Water at 20°C
Formula Weight 119.37
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Chloroform, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, contains amylene, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Chloroform
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.