Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, ACS Reagent, With Stabilizer (Amylene), ≥99.8%, Solstice

Catalog No. 60046959 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
6 x 1 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-959 6 x 1 L Glass Bottle
60-046-956 1 L Glass Bottle
60-046-957 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-046-959 Supplier Solstice Supplier No. 4724766X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent, contains amylenes as stabilizer,≥99.8%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point -61.3°C
Density 1.492g/mL at 25°C, 1.48g/mL at 25°C
Boiling Point 61°C
Odor Sweet
Packaging Glass Bottle
Quantity 6 x 1 L
UN Number UN1888
Beilstein 1731042
IUPAC Name trichloromethane
Vapor Density 4.1 (vs air)
Vapor Pressure 160mmHg (20°C)
Formula Weight 119.38g/mol
Percent Purity ≥99.8%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Chloroform, Containsamylenes as stabilizer
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338-P311
DOTInformation 6.1/PGIII
RTECSNumber 200-663-8
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.