Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, ACS, Reagents

Catalog No. 01338047
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Glass Bottle
Safety Coated Glass
Steel Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-338-047 4 L Safety Coated Glass
01-338-053 1 L Safety Coated Glass
01-338-051 2.5 L Safety Coated Glass
01-338-049 20 L Steel Pail
01-338-048 4 L Glass Bottle
01-338-046 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-338-047 Supplier Reagents Holdings Llc Supplier No. C11755004C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chloroform, ACS, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym Trichloromethane, Methyl trichloride
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.5°C
Color Colorless
Quantity 4 L
Packaging Safety Coated Glass
Physical Form Liquid
Specific Gravity 1.484
Boiling Point 60.5°C
Grade ACS
Chemical Name or Material Chloroform, ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.