Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, for HPLC, GC, pesticide residue analysis, and spectrophotometry, contains amylene, ≥99.9%, Solstice

Catalog No. 6001744 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
HDPE Nowpak™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-017-44 20 L HDPE Nowpak™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-017-44 Supplier Solstice Supplier No. NP04920

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-66-3 , 513-35-9
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

CAS Max % 99.9900%
Density 1.49 g/cm3
Quantity 20 L
Packaging HDPE Nowpak™
UN Number UN1888
Boiling Point 62°C
Formula Weight 119.37
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Chloroform, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, contains amylene, ≥99.9%
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Chloroform
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.