Learn More
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 83.754, 15.4, 0.846 |
| Decomposition Information | Hydrogen chloride; Carbon monoxide; Carbon dioxide |
| Density | 1.3g/mL |
| Identification | Passes Test |
| Packaging | Amber Glass |
| Physical Form | Liquid |
| Specific Gravity | 1.3g/mL |
| Linear Formula | CHCl3 |
| UN Number | UN2810 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Causes damage to organs (liver, kidneys, central nervous system, hearing organ) through prolonged or repeated exposure (inhalation, oral). Toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Do not breathe mist, spray, vapors. Wear eye protection, face protection, protective clothing, protective gloves. Wash exposed skin thoroughly after handling. Do not eat, drink or smoke when using this product. Avoid release to the environment. If exposed or concerned: Get medical advice/attention. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. If on skin: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.