Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, CHROMASOLV™ for residue analysis, contains ∼1% ethanol as stabilizer, ≥99.8%, Solstice

Catalog No. 60048237 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-237 4 x 4 L Glass Bottle
60-045-711 4 x 2.5 L Glass Bottle
60-017-43 1 L Glass Bottle
60-045-710 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-048-237 Supplier Solstice Supplier No. 256694X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™ for residue analysis, contains ∼1% ethanol as stabilizer,≥99.8%

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3 , 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C (literature)
CAS Min % ≥90.0000%, ≥0.1000%
CAS Max % <1.0000%
Density 1.492 g/mL at 25 °C(lit.)
Quantity 4 x 4 L
Packaging Glass Bottle
Linear Formula CHCl3
UN Number UN1888
Beilstein 1731042
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride
Boiling Point 60°C to 62°C
Percent Purity >99.8%
Grade Residue Analysis
Assay GC product+ethanol
Chemical Name or Material Chloroform, CHROMASOLV™ for residue analysis, contains ∼1% ethanol as stabilizer, ≥99.8%
Show More Show Less
ShelfLife 450 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Chloroform
EINECSNumber 200-663-8
Recommended Storage Room Temp
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.