Learn More
Chloroform - D1, 0.03 vol.% TMS, deuteration deg. min. 99.8% for NMR, MagniSolv™, MilliporeSigma™
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Physical Form | Liquid |
| Color | Colorless |
| Quantity | 25 mL |
| Packaging | Glass Bottle |
| Odor | Characteristic |
| Vapor Pressure | 211 hPa at 20°C (68°F) |
| Assay Percent Range | 99.8% min. |
| Formula Weight | 120.38 |
| Chemical Name or Material | Chloroform - D1, 0.03 vol.% TMS, Stabilized with silver |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.