Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, Solstice

Catalog No. 60090174 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
Packaging:
Returnable Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-174 56 L Returnable Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-174 Supplier Solstice Supplier No. CS04856
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For HPLC, GC, pesticide residue analysis, and spectrophotometry, contains amylene and 1% ethanol, >99.9%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
PubChem CID 6212
ChEBI CHEBI:35255

Specifications

Acidity 1 mg/L max.
Quantity 56 L
Packaging Returnable Steel Drum
Physical Form Liquid, Clear
Refractive Index 1.4443 - 1.4451
Recommended Storage Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.