Learn More
Description
- Chloroform HPLC Solvent and Reagent Amylene Stabilized, Safety: Toxic
Specifications
Specifications
| Color | Colorless |
| Appearance | Colorless Liquid |
| Density | 1.484 g/cm3 |
| Quantity | 4 L |
| Odor | Characteristic, pleasant, sweet |
| Packaging | Amber Glass |
| Physical Form | Liquid |
| Residue after Evaporation | <3 ppm |
| Specific Gravity | 1.484 |
| Vapor Pressure | 211 hPa at 20 °C |
| Show More |
Safety and Handling
| Hazard Category | Acute toxicity |
| Hazard Statement | Carcinogenicity |
| Precautionary Statement | Skin Irritation |
| ShelfLife | 3 Years |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.