Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, HPLC Grade, (Ethanol Stabilized), MilliporeSigma™

Catalog No. MCX1058P1 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MCX1058P1 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MCX1058P1 Supplier MilliporeSigma Supplier No. CX1058P1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Density 1.465-1.476g/ml
Quantity 4 L
Packaging Glass Bottle
Physical Form Clear liquid, free from particulates
Residue after Evaporation 2ppm
Vapor Pressure 211 hPa (20°C)
UN Number UN 1888
Water 0.01%
Boiling Point 61°C (1013 hPa)
Solubility Information 8 g/L in water
Formula Weight 119.38
Percent Purity 99.8%
Chemical Name or Material Chloroform, Stabilized with 0.75% ethanol.
Show More Show Less
Hazard Statement H302 Harmful if swallowed. H315 Causes skin irritation. H319 Causes serious eye irritation. H331 Toxic if inhaled. H351 Suspected of causing cancer. H361 Suspected of damaging fertility or the unborn child. H372 Causes damage to organs (Liver, Kidney) through prolonged or repeated exposure
Precautionary Statement P201 Obtain special instructions before use. P202 Do not handle until all safety precautions have been read and understood. P260 Do not breathe dust/ fume/ gas/ mist/ vapors/ spray. P264 Wash skin thoroughly after handling. P270 Do not eat, drink or smoke when using this product. P271 Use only outdoors or in a well-ventilated area. P280 Wear protective gloves/ eye protection/ face protection. P302 + P352 IF ON SKIN: Wash with plenty of soap and water. P304 + P340 IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 200-663-8
RTECSNumber FS9100000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.