Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform Isoamyl Alcohol Mixture BioUltra, 24:1, ≥99.5% (chloroform + isoamyl alcohol, GC), MilliporeSigma™ Supelco™

Catalog No. 111016907 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6907 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6907 Supplier MilliporeSigma Supelco Supplier No. 25666100ML
Add to Cart
Add to Cart

Specifications

CAS 67-66-3
MDL Number MFCD00171265
Type Standard
Quantity 100 mL
Percent Purity ≥99.5% (chloroform + isoamyl alcohol, GC)
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.