Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, MP Biomedicals™

Catalog No. MP219381404 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219381404 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219381404 Supplier MP Biomedicals, Inc Supplier No. 0219381404
Add to Cart
Add to Cart

Chloroform is typically used as a solvent and as a cleansing agent.

Procedures have been described for the use of chloroform in lambda plaques storage, lambda cDNA storage, the removal of mineral oil from PCR reaction samples, oligonucleotide purification, a hydroxyl radical footprinting protocol, a transcriptional run-on assay protocol, and an overlay assay for beta-galactosidase activity.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Quantity 4 L
Physical Form Liquid
Assay Percent Range ≥99.8%
Solubility Information Soluble in water (1mL/200mL water); miscible with ethanol, benzene, ether, petroleum ether, carbon tetrachloride, carbon disulfide, oils.
Formula Weight 119.4g/mol
Grade ACS
Recommended Storage Store at Room Temperature (15–30°C). Protect from light.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.