Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, OmniPur™,, MilliporeSigma™

Catalog No. 3150950ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
950 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
31-509-50ML 950 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-509-50ML Supplier MilliporeSigma Supplier No. 3150950ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane

Specifications

Melting Point -63°C
Color Colorless
Quantity 950 mL
Odor Pleasant. Ethereal
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.49
Vapor Pressure 22.3 kPa (167mm Hg) [20°C]
UN Number 1888
Boiling Point 60.5°C
Solubility Information Partially soluble in water
Formula Weight 119.38
Percent Purity ≥99.8%
Chemical Name or Material Chloroform
Show More Show Less
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containersthat have been opened must be carefully resealed and kept upright to prevent leakage.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.