Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChloroForm, Reagent grade, For ACS analysis, ≥99.8%, MP Biomedicals™

Catalog No. ICN19381491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.5 L
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19381491 1 L
ICN19381490 0.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19381491 Supplier MP Biomedicals, Inc Supplier No. 0219381491
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Density 1.476 to 1.478 g/cm3 at 20°C
Quantity 1 L
Physical Form Liquid
Assay Percent Range ≥99.8%
IUPAC Name chloroform
Formula Weight 119.4
Grade Reagent
Chemical Name or Material Chloroform, For ACS analysis
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.