Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, ReagentPlus™, ≥99.8%, Solstice

Catalog No. 60044791 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-791 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-791 Supplier Solstice Supplier No. 1329502.5L

missing translation for 'validMainframeMsg'

Reagent Grade, contains 0.5-1.0% ethanol as stabilizer,≥99.8%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Density 1.492 g/mL at 25°C, 1.48 g/mL at 25°C
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.445(lit.)
Vapor Pressure 160 mmHg (20°C)
UN Number UN1888
Beilstein 1731042
Boiling Point 60°C to 62°C
Formula Weight 119.38g/mol
Percent Purity ≥99.8%
Grade ReagentPlus
Chemical Name or Material Chloroform
Show More Show Less
Hazard Statement P260-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338-P403 + P233
Precautionary Statement H302-H315-H319-H331-H336-H351-H361d-H372
DOTInformation 6.1/PGIII
EINECSNumber 200-663-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.