Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, stabilized with ethanol, for HPLC, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
67-66-3,64-17-5
CHCl3
119.369
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
67-66-3,64-17-5
CHCl3
119.369
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
For HPLC
Absorbance 0.009 max. at 300nm,0.046 max. at 265nm,0.301 max. at 250nm,0.699 max. at 245nm,1 cm cell vs water,1 cm cell vs water at 0nm
Absorbance Conditions 1 cm cell vs water
Assay Percent Range 99+%
Linear Formula CHCl3
Merck Index 15, 2142
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
Formula Weight 119.38
Physical Form Crystals
Percent Purity ≥99%
Grade HPLC
Density 1.4800g/mL
Free Acid 0.001 % max. (HCl)
Free Chlorine 0.00003 % max. (as Cl)
Packaging Glass bottle
Refractive Index 1.4443 to 1.4463
Residue after Evaporation 0.0002 % max.
Solubility Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
Specific Gravity 1.48
Stabilizer 0.5 to 1.0 % ethanol
Water 0.01% Max. (K.F.)
Viscosity 0.56 mPa.s (20°C)
Boiling Point 61.0°C
Color White
Melting Point -63.0°C
Quantity 1L
Chemical Name or Material Chloroform, Stabilized with ethanol
Show More Show Less

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning