Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, Stabilized, Reagent, ACS, 99.8%, Spectrum™ Chemical
SDP

Catalog No. 18601612 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-612 500 mL Amber Glass Bottle
18-601-611 100 mL Amber Glass Bottle
18-601-614 1 L Amber Glass Bottle
18-601-613 6 x 500 mL Amber Glass Bottle
18-601-615 4 L Amber Glass Bottle
18-601-616 6 x 1 L Amber Glass Bottle
18-601-617 4 x 4 L Amber Glass Bottle
18-601-618 20 L Steel Can
18-601-619 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18-601-612 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1220500MLGL
Add to Cart
Add to Cart

C1220, 67-66-3, CHCl3

Spectrum™ Chemical Chloroform, Stabilized, Reagent, ACS , is a colorless, sweet smelling organic compound comes in the form of a dense liquid and is a trihalomethane. It is also commonly used as laboratory solvent because it is relatively unreactive, conveniently volatile and is miscible with most organic compounds. Although it uses as a reagent are limited, it is used in organic synthesis in order to produce dichlorocarbene by reacting with sodium hydroxide. As an ACS grade Reagent, Spectrum Chemical manufactured chloroform is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

CAS Max % 100%
Quantity 500 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.001%
UN Number UN1888
Boiling Point 61°C
Percent Purity 99.8%
Grade Reagent
Chemical Name or Material Chloroform
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.