Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroiodomethane, 98%, Thermo Scientific™

Catalog No. AC222150100
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC222150100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC222150100 Supplier Thermo Scientific Chemicals Supplier No. 222150100

missing translation for 'validMainframeMsg'

Alkylation reagent

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-71-5 , 7440-22-4
Molecular Formula CH2ClI
Molecular Weight (g/mol) 176.381
InChI Key PJGJQVRXEUVAFT-UHFFFAOYSA-N
Synonym chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
PubChem CID 11644
IUPAC Name chloro(iodo)methane
SMILES C(Cl)I

Specifications

Density 2.4200g/mL
Boiling Point 108.0°C to 109.0°C
Refractive Index 1.5812 to 1.5832
Linear Formula ClCH2I
Quantity 10 mL
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in many organic solvents
Formula Weight 176.38
Chemical Name or Material Chloroiodomethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.