Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyl Chloroformate 95.0+%, TCI America™
SDP

Catalog No. C1396100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1396100G 100 g
C139625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1396100G Supplier TCI America Supplier No. C1396100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22128-62-7
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.936
MDL Number MFCD00077688
InChI Key JYWJULGYGOLCGW-UHFFFAOYSA-N
Synonym chloromethyl chloroformate, chloromethylchloroformate, chloroformic acid chloromethyl ester, carbonochloridic acid, chloromethyl ester, palite, chlormethylchlorformiat, clco2ch2cl, chlormethylchlorocarbonat, chloromethyl choroformate, choromethyl chloroformate
PubChem CID 62754
IUPAC Name chloromethyl carbonochloridate
SMILES C(OC(=O)Cl)Cl

Specifications

Color Yellow
Boiling Point 108°C
Quantity 100 g
UN Number 2745
Formula Weight 128.94
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Chloromethyl Chloroformate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.