Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Chloromethyl)cyclopropane 95.0+%, TCI America™
SDP

Catalog No. C246925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C246925G 25 g
C24695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C246925G Supplier TCI America Supplier No. C246925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5911-08-0
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00001307
InChI Key ZVTQWXCKQTUVPY-UHFFFAOYSA-N
Synonym chloromethyl cyclopropane, cylopropylmethyl chloride, cyclopropylmethyl chloride, cyclopropane, chloromethyl, chloromethyl-cyclopropane, j7okqcc0kw, unii-j7okqcc0kw, cylopropylmethylchloride, chlorocyclopropylmethane, cyclopropylchloromethane
PubChem CID 80013
IUPAC Name (chloromethyl)cyclopropane
SMILES ClCC1CC1

Specifications

Melting Point -91°C
Color Yellow
Boiling Point 88°C
Quantity 25 g
UN Number 1993
Formula Weight 90.55
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (Chloromethyl)cyclopropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.