Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyl Isopropyl Carbonate 98.0+%, TCI America™
SDP

Catalog No. C2626100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2626100G 100 g
C262625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2626100G Supplier TCI America Supplier No. C2626100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35180-01-9
Molecular Formula C5H9ClO3
Molecular Weight (g/mol) 152.57
MDL Number MFCD07375443
InChI Key JHYNXXBAHWPABC-UHFFFAOYSA-N
Synonym Carbonic Acid Chloromethyl Isopropyl Ester
PubChem CID 14787051
IUPAC Name chloromethyl propan-2-yl carbonate
SMILES CC(C)OC(=O)OCCl

Specifications

Color Yellow
Quantity 100 g
UN Number 2924
Formula Weight 152.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethyl Isopropyl Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.