Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyl Methyl Ether Solution, 1000μg/mL in Methanol, Ultra Scientific

Catalog No. USEPA1209
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1209 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1209 Supplier Agilent Technologies Supplier No. EPA12091
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107-30-2 , 67-56-1
Molecular Formula C2H5ClO
Molecular Weight (g/mol) 80.511
InChI Key XJUZRXYOEPSWMB-UHFFFAOYSA-N
PubChem CID 7864
ChEBI CHEBI:82271
IUPAC Name chloro(methoxy)methane
SMILES COCCl

Specifications

Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Chloromethyl Methyl Ether, 1000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.