Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Chloromethyl)triphenylphosphonium Chloride 98.0+%, TCI America™
SDP

Catalog No. C100925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C100925G 25 g
C10095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C100925G Supplier TCI America Supplier No. C100925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5293-84-5
Molecular Formula C19H17Cl2P
Molecular Weight (g/mol) 347.22
MDL Number MFCD00011867
InChI Key SXYFAZGVNNYGJQ-UHFFFAOYSA-M
Synonym chloromethyl triphenylphosphonium chloride, chloromethyltriphenylphosphonium chloride, chloromethyl triphenylphosphoniumchloride, phosphonium, chloromethyltriphenyl-, chloride, chloromethly triphenylphosphonium chloride, chloromethyl triphenylphosphanium chloride, chloromethyl triphenylphosphonium chliride, triphenyl chloromethyl-phosphonium chloride, acmc-1albk, phosphonium, chloromethyl triphenyl-, chloride
PubChem CID 197823
IUPAC Name (chloromethyl)triphenylphosphanium chloride
SMILES [Cl-].ClC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 347.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (Chloromethyl)triphenylphosphonium Chloride
RTECSNumber TA1882000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.