Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyldimethylphenylsilane 97.0+%, TCI America™
SDP

Catalog No. C141925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C141925G 25 g
C14195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C141925G Supplier TCI America Supplier No. C141925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1833-51-8
Molecular Formula C9H13ClSi
Molecular Weight (g/mol) 184.74
MDL Number MFCD00040858
InChI Key RJCTVFQQNCNBHG-UHFFFAOYSA-N
PubChem CID 74598
IUPAC Name (chloromethyl)dimethylphenylsilane
SMILES C[Si](C)(CCl)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 225°C
Quantity 25 g
Formula Weight 184.74
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethyldimethylphenylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.