Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Chloromethylene)dimethyliminium Chloride 95.0+%, TCI America™
SDP

Catalog No. C1545250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1545250G 250 g
C154525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1545250G Supplier TCI America Supplier No. C1545250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3724-43-4
Molecular Formula C3H7Cl2N
Molecular Weight (g/mol) 127.996
MDL Number MFCD00011868
InChI Key QQVDYSUDFZZPSU-UHFFFAOYSA-M
Synonym arnold's reagent, vilsmeier reagent, chloromethylene dimethyliminium chloride, methanaminium, n-chloromethylene-n-methyl-, chloride, chloromethylene dimethylammonium chloride, n-chloromethylidene-n-methylmethanaminium chloride, chloromethylenedimethylammonium chloride, chloromethylidene dimethylazanium chloride, n-chloromethylene-n,n-dimethyl ammonium chloride
PubChem CID 77311
IUPAC Name chloromethylidene(dimethyl)azanium;chloride
SMILES C[N+](=CCl)C.[Cl-]

Specifications

Quantity 250 g
UN Number 2923
Formula Weight 128.00
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Chloromethylene)dimethyliminium Chloride
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.