Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroquine Diphosphate 98.0+%, TCI America™
SDP

Catalog No. C2301100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2301100G 100 g
C230125G 25 g
C2301500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C2301100G Supplier TCI America Supplier No. C2301100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50-63-5
Molecular Formula C18H32ClN3O8P2
Molecular Weight (g/mol) 515.86
MDL Number MFCD00069852
InChI Key QKICWELGRMTQCR-UHFFFAOYNA-N
Synonym chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine
PubChem CID 64927
IUPAC Name 7-chloro-N-[5-(diethylamino)pentan-2-yl]quinolin-4-amine; bis(phosphoric acid)
SMILES OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 100 g
Formula Weight 515.86
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chloroquine Diphosphate
EINECSNumber (9)-0279
RTECSNumber VB2450000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.