Learn More
Chlorosulfonic acid, typically 99%
Description
Chlorosulfonic acid is used as detergent and as an anti-contrail agent. It is used as an intermediate in the production of other substances such as pharmaceuticals and chemicals. It finds application in producing smoke screens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Appearance | Yellow to brown liquid |
| Color | Yellow to Brown |
| Melting Point | -80°C |
| Boiling Point | 158°C |
| Density | 1.753 g/cm3 at 20°C |
| Odor | Odorless |
| Assay Percent Range | typically 99% |
| Quantity | 1 kg |
| Physical Form | Liquid |
| UN Number | 1754 |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | DOT Class: 8, Packaging Group: I |
| EINECSNumber | 232-234-6 |
| RTECSNumber | FX5730000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.