Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chlorothalonil Monoclonal Antibody (4D7)
GREENER_CHOICE

Catalog No. PIMA549715
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549715 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549715 Supplier Invitrogen™ Supplier No. MA549715
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Chlorothalonil is a broad-spectrum fungicide that is commonly used in agriculture to control fungal diseases on crops such as vegetables, fruits, and ornamental plants. It is also used in residential settings to control mold and mildew on surfaces such as wood, concrete, and paint. Chlorothalonil works by inhibiting the growth and reproduction of fungi, preventing them from causing damage to plants or surfaces.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chlorothalonil
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 4D7
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of CTL hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.