Learn More
Strem, An Ascensus Company CAS# 20717-86-6. 25g. Chlorotitanium triisopropoxide, 97%. MFCD00009861
SDP
Supplier:  Strem, An Ascensus Company 22038025G
Encompass
CAS# 20717-86-6. 25g. Chlorotitanium triisopropoxide, 97%. MFCD00009861. Molecular Weight: 260.62. Molecular Formula: TiCl(OC3H7)3. Color/form: fused solid to pale yellow liq. Strem# 22-0380. http://www.strem.com/catalog/v/22-0380/
Catalog No. 50-901-10739
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.