Learn More
Sigma Organic Chemistry Chlorotrimethylsilane | 500ML | 75-77-4 | MFCD00000502
Supplier: Sigma Organic Chemistry 92360500ML

Chlorotrimethylsilane, 500ML
About This Item:
Synonym(s): TMCS; Trimethylchlorosilane; Trimethylsilyl chloride
Linear Formula: (CH3)3SiCl
Molecular Weight: 108.64
CAS Number: 75-77-4
MDL Number: MFCD00000502
UNSPSC Code: 12352101
Purity: >=99.0% (GC)
Boiling Point: 57 C (lit.)
Melting Point: -40 C (lit.)
Density: 0.856 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.387 (lit.)
Storage: room temp
EINECS Number: 200-900-5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.