Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chlorotripropylsilane, 98%

Catalog No. p-4258909
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313930050 5 g Glass bottle
AC313930250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC313930050 Supplier Thermo Scientific Chemicals Supplier No. 313930050
Only null left
Add to Cart

CAS: 995-25-5 | C9H21ClSi | 192.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 995-25-5
Molecular Formula C9H21ClSi
Molecular Weight (g/mol) 192.81
InChI Key ACTAPAGNZPZLEF-UHFFFAOYSA-N
Synonym tri-n-propylchlorosilane, tripropylchlorosilane, silane, chlorotripropyl, chlorotri-n-propylsilane, chloro tripropyl silane, chloro-tri-n-propylsilane, chlorotripropylsilane, chloro tri-n-propyl silane
PubChem CID 70445
IUPAC Name chloro(tripropyl)silane
SMILES CCC[Si](CCC)(CCC)Cl

Specifications

Color Colorless
Density 0.8820g/mL
Boiling Point 199.0°C to 200.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3990 to 1.4410 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.882
Formula Weight 192.81
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material chlorotripropylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Store in a dry place. Store in a closed container.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Protect from moisture.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-624-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.