Learn More
Cayman Chemical Chlorpromazine (hydrochloride), 69-09-0, 1 g
SDP
Supplier:  Cayman Chemical 161291
Encompass_Preferred
A typical antipsychotic; an antagonist of dopamine D2, D3, and D4 receptors (Kis = 0.66, 0.84, and 1.2 nM, respectively) as well as 5-HT2A receptors (Ki = 1.8 nM); an antagonist of histamine H1, α1a-, α2b-, and α2C-adrenergic, and M3 muscarinic acetylcholine receptors (Kis = 6, 0.28, 27, 46, and 47 nM, respectively); increases latency to find the platform in a repealed acquisition water maze task and decreases vertical activity and stereotypic movements in the open field test in rats at 10 mg/kg; reduces cisplatin-induced emesis in dogs at 0.3, 1, and 3 mg/kg
Catalog No. NC1264063
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.