Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorpromazine hydrochloride, 98+%

Catalog No. AAJ6365914
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6365914 25 g
AAJ6365909 10 g
AAJ6365922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6365914 Supplier Thermo Scientific Chemicals Supplier No. J6365914
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 69-09-0 | C17H20Cl2N2S | 355.321 g/mol

Chlorpromazine hydrochloride is used as a substitute for benzidine, o-dianisidine and o-tolidine in the determination of microquantities of hemoglobin and peroxidase. Chlorpromazine, Hydrochloride is used in cell signaling and neuroscience research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorpromazine hydrochloride is used as a substitute for benzidine, o-dianisidine and o-tolidine in the determination of microquantities of hemoglobin and peroxidase. Chlorpromazine, Hydrochloride is used in cell signaling and neuroscience research.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-09-0
Molecular Formula C17H20Cl2N2S
Molecular Weight (g/mol) 355.321
MDL Number MFCD00012654
InChI Key FBSMERQALIEGJT-UHFFFAOYSA-N
Synonym chlorpromazine hydrochloride, sonazine, chlorpromazine hcl, chloractil, klorpromex, promacid, hebanil, propaphenin, hibernal, hybernal
PubChem CID 6240
ChEBI CHEBI:3649
IUPAC Name 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2SC3=C1C=C(C=C3)Cl.Cl

Specifications

Melting Point 196°C
Color White
Quantity 25 g
UN Number UN2811
Beilstein 3779989
Merck Index 14,2185
Solubility Information Soluble in water
Formula Weight 355.3
Percent Purity ≥98%
Physical Form Crystalline Solid
Chemical Name or Material Chlorpromazine hydrochloride
Show More Show Less
Hazard Category H301-H330
Hazard Statement GHS H Statement
H301-H330
Toxic if swallowed.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-701-3
RTECSNumber SO1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.