Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorpromazine Hydrochloride, MP Biomedicals™

Catalog No. MP219032690 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219032690 500 g
MP219032691 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219032690 Supplier MP Biomedicals, Inc Supplier No. 0219032690
Add to Cart
Add to Cart

Inhibits calmodulin-dependent stimulation. Chlorpromazine Hydrochloride, MP Biomedicals is a phenothiazine antipsychotic and a D2 and H1 antagonist. It is a substitute for benzidine, o-dianisidine, and o-tolidine in the determination of microquantities of

Chlorpromazine Hydrochloride inhibits calmodulin-dependent stimulation of cyclic nucleotide phosphodiesterase and nitric oxide synthase, and it is reported to uncouple and inhibit oxidative phosphorylation at 10-3 – 10-4 M.

  • Demonstrates cytotoxic and antiproliferative activity against leukemic cells, but does not affect the viability of normal lymphocytes
  • Phenothiazine antipsychotic
  • D2 dopamine and H1 histamine receptors antagonist
  • Possibly a photo-genotoxic agent in humans
  • Helps to induce hypothermia in mice in the dose range of 12.5–100mg/kg
  • A substitute for benzidine, o-dianisidine, and o-tolidine in the determination of microquantities of hemoglobin and peroxidase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-09-0
Molecular Formula C17H20Cl2N2S
Molecular Weight (g/mol) 355.321
InChI Key FBSMERQALIEGJT-UHFFFAOYSA-N
Synonym chlorpromazine hydrochloride, sonazine, chlorpromazine hcl, chloractil, klorpromex, promacid, hebanil, propaphenin, hibernal, hybernal
PubChem CID 6240
ChEBI CHEBI:3649
IUPAC Name 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2SC3=C1C=C(C=C3)Cl.Cl

Specifications

Melting Point 194°C to 199°C
Loss on Drying ≤1.0%
Quantity 500 g
Solubility Information Soluble in water (50mg/mL - clear, colorless to faint yellow-grey solution), methanol, ethanol, chloroform; practically insoluble in ether, benzene.
Formula Weight 355.3g/mol
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.