Learn More
Crescent Chemical Co Inc CHLORPYRIFOS D10 20 MG
SDP
Supplier:  Crescent Chemical Co Inc PSC11600100
Encompass_Preferred
Chlorpyrifos D10 Insecticide (Isotop neat) CAS 285138-81-0 Molecular Weight - 360.6479 Storage - -18°C Catalog Num - DRE-C11600100 pesticide
Catalog No. 50-730-59
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.