Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorthalidone, 98%

Catalog No. p-7077862
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460932500 250 mg
AC460930010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC460932500 Supplier Thermo Scientific Chemicals Supplier No. 460932500
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 77-36-1 | C14H11ClN2O4S | 338.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-36-1
Molecular Formula C14H11ClN2O4S
Molecular Weight (g/mol) 338.77
InChI Key JIVPVXMEBJLZRO-UHFFFAOYSA-N
Synonym chlorthalidone, chlortalidone, phthalamudine, chlorphthalidolone, hygroton, chlorthalidon, phthalamodine, chlorothalidone, chlorphthalidone, natriuran
PubChem CID 2732
ChEBI CHEBI:3654
IUPAC Name 2-chloro-5-(1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide
SMILES C1=CC=C2C(=C1)C(=O)NC2(C3=CC(=C(C=C3)Cl)S(=O)(=O)N)O

Specifications

Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Formula Weight 338.77
Percent Purity 98%
Chemical Name or Material Chlorthalidone
EINECSNumber 201-022-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.