Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorzoxazone 98.0+%, TCI America™
SDP

Catalog No. C2237250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2237250G 250 g
C223725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2237250G Supplier TCI America Supplier No. C2237250G

Chemical Identifiers

CAS 95-25-0
Molecular Formula C7H4ClNO2
Molecular Weight (g/mol) 169.564
MDL Number MFCD00005717
InChI Key TZFWDZFKRBELIQ-UHFFFAOYSA-N
Synonym chlorzoxazone, chlorzoxazon, paraflex, chloroxazone, 5-chloro-2-benzoxazolone, biomioran, escoflex, myoflexin, myoflexine, pathorysin
PubChem CID 2733
ChEBI CHEBI:3655
IUPAC Name 5-chloro-3H-1,3-benzoxazol-2-one
SMILES C1=CC2=C(C=C1Cl)NC(=O)O2

Specifications

Melting Point 191°C
Quantity 250 g
Formula Weight 169.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorzoxazone
RTECSNumber DM5250000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.