Learn More
Cygnus Technologies Cygnus Technologies CHO HCP 3G Assay Reagent Set, High-Throughput Immunoassay, Semi-Quantitative Detection, unit: Set
Supplier:  Cygnus Technologies G5501
Encompass
The CYGNUS TECHNOLOGIES CHO HCP 3G Assay Reagent Set is a semi-quantitative immunoassay designed for detecting host cell protein (HCP) contamination in products from CHO cell lines. It offers rapid, high-throughput analysis with integration for Gyrolab systems, ensuring streamlined purification process development and quality control for biopharmaceuticals. This reagent set stands out due to its broad specificity and robust methodology, making it essential for routine quality assurance and product release.
Catalog No. NC2187818
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.