Learn More
Genscript Corporation CHO-K1/CXCR4/G15 Stable Cell
Supplier: Genscript Corporation M00556
CXCR4 is a receptor for the C-X-C chemokine SDF-1 (Stromal Cell-Derived Factor 1) It is involved in haematopoiesis and cardiac ventricular septum formation and plays an essential role in vascularization of the gastrointestinal tract cerebellar development and survival of hippocampal-neuron of central nerve system CXCR4 also acts as a primary receptor for some HIV-2 isolates and as a co-receptor with CD4 for HIV-1 X4 viruses
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.