Learn More
Genscript Corporation cTnT 26D7 mAb Mouse
Supplier:  Genscript Corporation V004021
Encompass_Preferred
Cardiac TnT (cTnT) is the largest subunit of the troponin complex which is composed of cTnT troponin I (TnI) troponin C (TnC) It is encoded by the TNNT2 gene with a calculated molecular weight of 34 6 kDa The cTnT is considered as a useful biomarker for the diagnosis of acute myocardial infarction
Catalog No. 50-000-56742
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.