Learn More
Genscript Corporation CHO-K1/IP1/G15 Stable Cell Li
Supplier:  Genscript Corporation M00310
Encompass_Preferred
IP1 receptor is a receptor for prostaglandin I2 or prostacyclin When binding with a prostacyclin molecule the receptor changes conformation and activates Gs Prostaglandin I2 the major product of cyclooxygenase in macrovascular endothelium mediates a potent vasodilation and inhibition of platelet aggregation by binding to this receptor
Catalog No. 50-000-57393
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.