Learn More
Genscript Corporation CHO-K1/ADRA1D Stable Cell Line
Supplier: Genscript Corporation M00340
Recombinant CHO-K1 cells stably overexpress human adrenoceptor alpha 1D (ADRA1D) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.