Learn More
Genscript Corporation CHO-K1/PTH1/G15 Stable Cell L
Supplier:  Genscript Corporation M00315
Encompass_Preferred
Recombinant CHO-K1 cells stably overexpress human parathyroid hormone 1 receptor (PTH1) on the surface and contain high levels of G protein G s and G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57392
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.