Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Chocolate Agar Medium
GSA_VA

Catalog No. r01300
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
150/Cs.
20/Pk.
Format:
10mm x 100mm Spacesaver Plate
85mm Monoplate
D - slant
Media Type:
Chocolate Agar Medium
Chocolate Agar; Enriched
Manufacturing Origin:
United States
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Manufacturing Origin
R01300 10/Pk. 85mm Monoplate Chocolate Agar Medium United States
R01302 100/Pk. 85mm Monoplate Chocolate Agar Medium United States
R060482 20/Pk. D - slant Chocolate Agar; Enriched United States
R01293 150/Cs. 10mm x 100mm Spacesaver Plate Chocolate Agar; Enriched United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R01300 Supplier Thermo Scientific™ Supplier No. R01300
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Obtain fast and accurate results with a nutritious medium for the isolation of fastidious organisms Neisseria, Haemophilus, and Gardnerella vaginalis.

Cultivate and isolate fastidious organisms especially Neisseria spp., Haemophilus, and Gardnerella vaginalis using Thermo Scentific™ Chocolate Agar.

Hemoglobin in the Chocolate Agar formulation provides the X factor, and GCHI Enrichment provides the V factor required for growth of fastidious organisms. The presence of starch ensures that toxic metabolites produced by Neisseria are absorbed, and phosphate buffers are included to prevent changes in pH due to amine production that would affect the survival of the organism.

Neisseria are capnophilic, gram-negative diplococci or rods that occur in pairs or short chains and inhabit the mucous membranes of humans and animals. Neisseria gonorrhoeae and Neisseria meningitidis are two important pathogens in this group. Gonorrhea is a sexually transmitted disease caused by N. gonorrhoeae and is one of the most frequently reported infectious diseases in the United States and worldwide. This organism is oxidase-positive, and growth is best obtained on Chocolate Agar or Thayer Martin Agar. The treatment and control of gonorrhea is complicated by increasing resistance to a variety of antimicrobial agents, including penicillin, tetracycline, and fluoroquinolones.

Haemophilus
are small, pleomorphic, gram-negative coccobacilli. They are nonmotile, non-sporeforming, fastidious, facultative anaerobes. Almost all species require preformed growth factors present in the blood, particularly the X factor (hemin) and/or V factor (NAD) for growth.

Gardnerella vaginalis is found in the genitourinary tract of females and is an indicator organism for bacterial vaginosis (BV). BV is a risk factor for premature labor, chorioamnionitis, and postpartum endometritis. Women with postpartum fever and neonates with sepsis may be infected with Gardnerella vaginalis. In males, Gardnerella vaginalis has been associated with urinary tract infections, prostatitis, and urethritis. Gardnerella vaginalis is a gram-variable rod or coccobacillus. Beta hemolysis is observed on human or rabbit blood-containing media, but not on Sheep Blood Agar.

Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Opaque brown
Content And Storage 2°C to 8°C
Format 85mm Monoplate
Manufacturing Origin United States
Media Type Chocolate Agar Medium
Product Type Agar Plate
Fill Volume 18.5 mL
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.