Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ CHOgro™ Complex Formation Solution

Catalog No. MIR6210 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6210 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR6210 Supplier Mirus Bio™ Supplier No. MIR 6210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHOgro™ Complex Formation Solution is an animal-origin-free solution specifically formulated for optimal transfection-complex formation in conjunction with CHOgro™ Expression Media and TransIT-PRO™ Transfection Reagent.

  • CHOgro™ Complex Formation Solution is an animal-origin-free solution specifically formulated for optimal transfection-complex formation in conjunction with CHOgro™ Expression Media and TransIT-PRO™ Transfection Reagent to provide a completely animal-origin-free expression system.
  • Manufactured under cGMP conditions in an ISO-compliant facility.

Specifications

Content And Storage Store at 4°C
Cell Type CHO
Format Bottle
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Transfection
Product Type Transfection Complex Formation Solution
Quantity 100 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.