Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ CHOgro™ Expression System

Catalog No. MIR6260 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6260 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR6260 Supplier Mirus Bio™ Supplier No. MIR 6260
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The CHOgro™ Expression System is a high-titer transient transfection system for suspension CHO cells.

The CHOgro Expression System was developed through systematic optimization of transfection protocol parameters including: cell density, transfection reagent, media formulation and culture temperature. With the CHOgro Expression System, high protein titers can now be achieved in suspension CHO cells through high density transient transfection.

  • Efficient: Enables high protein titers with simple workflow
  • Convenient: Quick adaptation to CHO cell lineages
  • Optimized: High-density growth with minimal cell clumping post transfection
  • Worry-free: No commercial license required; animal origin free

Specifications

Content And Storage Multiple storage conditions – see individual bottles for specific recommendations
Cell Type CHO
Transfection Technique Chemical Transfection
For Use With (Application) Transfection
Product Type Expression System
Quantity 1 Kit
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.